Learn More
Alkali Scientific Alkali Scientific™ 5L Roller Bottles (5000mL), 4250cm² Surface Area, Individually Wrapped, Sterile, Printed Graduations, 12/Cs, Non-Treated, Vent Cap (0.22µm hydrophobic membrane)
SDP
Catalog No. 503028174
Encompass_Preferred
Our 5000mL Roller Bottle is an essential tool for tissue culture applications. With printed graduations for easy volume measurement, this bottle simplifies the process. The vented cap ensures proper airflow while maintaining a sterile environment vital for cell culture growth. Each bottle comes sterile and ready for use, conveniently packed 12 per case. Ideal for laboratories, research facilities, and academic institutions, this roller bottle offers reliable performance and consistency in cell culture experiments.  Expanded Surface Roller Bottle increases surface area for cell culture Large knurled cap for easy grip Vented or non-vented cap for open or closed systems Manufactured from medical grade virgin polystyrene Tissue Culture Treated Roller Bottles have uniform hydrophilic surface ideal for consistent cell attachment Easy-to-read black printed graduations Packaged sterile in individual bags Vent caps feature 0.22µm hydrophobic membrane
Catalog No. 50-302-8174 Supplier Alkali Scientific Supplier No. VNT5100V
Our 5000mL Roller Bottle is an essential tool for tissue culture applications. With printed graduations for easy volume measurement, this bottle simplifies the process. The vented cap ensures proper airflow while maintaining a sterile environment vital for cell culture growth. Each bottle comes sterile and ready for use, conveniently packed 12 per case. Ideal for laboratories, research facilities, and academic institutions, this roller bottle offers reliable performance and consistency in cell culture experiments.  Expanded Surface Roller Bottle increases surface area for cell culture Large knurled cap for easy grip Vented or non-vented cap for open or closed systems Manufactured from medical grade virgin polystyrene Tissue Culture Treated Roller Bottles have uniform hydrophilic surface ideal for consistent cell attachment Easy-to-read black printed graduations Packaged sterile in individual bags Vent caps feature 0.22µm hydrophobic membrane