Learn More
AirSea USA 5L UN Steel Drum Lacquer Lined, 1A1/X1.6/600
Catalog No. 501534829
Encompass_Preferred
5L UN Steel drum with lacquer lining. UN Mark: 1A1/X1.6/600. This 5L steel drum is lacquer lined, includes a 51mm bung and is approved for shipping dangerous goods as liquids within packing groups I, II, III.
Catalog No. 50-153-4829 Supplier AirSea USA Supplier No. 230
5L UN Steel drum with lacquer lining. UN Mark: 1A1/X1.6/600. This 5L steel drum is lacquer lined, includes a 51mm bung and is approved for shipping dangerous goods as liquids within packing groups I, II, III.