Learn More
AirSea USA 5L UN Steel Drum Lacquer Lined, 1A1/X/550
Catalog No. 501534826
Encompass_Preferred
5L UN Steel Drum wth lacquer lining. UN Mark: 1A1/X/550. This 5L steel drum is lacquer lined, has a 51mm bung and is approved for shipping dangerous goods as liquids within packing groups I,II & III.
Catalog No. 50-153-4826 Supplier AirSea USA Supplier No. 112
5L UN Steel Drum wth lacquer lining. UN Mark: 1A1/X/550. This 5L steel drum is lacquer lined, has a 51mm bung and is approved for shipping dangerous goods as liquids within packing groups I,II & III.