Learn More
Andwin Scientific 5LEADER 1000 SINGLE CHANNEL P
SDP
Catalog No. 50820376
Encompass_Preferred
uleader; 1000 single channel pipettor, 100-1000 ul; Increments = 1ul
Catalog No. 50-820-376 Supplier Andwin Scientific Supplier No. 41101000
uleader; 1000 single channel pipettor, 100-1000 ul; Increments = 1ul