Learn More
Cayman Chemical 5-Lipoxygenase Polyclonal Antibody, 1 ea
SDP
Catalog No. 502973226
Encompass_Preferred
Immunogen: Synthetic peptide from an internal region of human 5-lipoxygenase • Host: Rabbit • Species Reactivity: (+) Human and porcine • Cross Reactivity: (–) 12-Lipoxygenase and 15-Lipoxygenase • Applications: WB and immunohistochemistry • MW = 78 kDa
Catalog No. 50-297-3226 Supplier Cayman Chemical Supplier No. 1604021 EA
Immunogen: Synthetic peptide from an internal region of human 5-lipoxygenase • Host: Rabbit • Species Reactivity: (+) Human and porcine • Cross Reactivity: (–) 12-Lipoxygenase and 15-Lipoxygenase • Applications: WB and immunohistochemistry • MW = 78 kDa