Learn More
ELKO FILTERING CO .5LON MESH 1 YRD 70 MICRON
SDP
Supplier:  ELKO FILTERING CO 0370/33
Encompass
Nylon Monofilament Precision Woven Mesh - 70 Micron Opening, Open Area 33%, 40 Inch wide, Sold by Yard - Available from 1 - 5000 Microns - Sefar 03-70/33
Catalog No. NC0446099