Learn More
Chemglass Life Sciences 5-Layer Cell Culture Flask, Vent Cap, Straight Neck, TC, Sterile, 1/pk, 8/cs
SDP
Catalog No. 501897650
Encompass_Preferred
Made from high clarity, 100% virgin polystyreneSterilized by E-beamVacuum plasma tissue culture treatmentClear lot number for batch traceabilityIndividually plastic wrappedNon-Pyrogenic, DNase/RNase-FreeHigh performance with 5 growth surfacesCell culture flasks have a growth area of 870cm². Tissue culture treated with a vent cap. Vent caps with 0.22μm hydrophobic filters for gas exchange. Recommended medium volume: 100-150mL.
Catalog No. 50-189-7650 Supplier Chemglass Life Sciences Supplier No. CGN2404V01
Made from high clarity, 100% virgin polystyreneSterilized by E-beamVacuum plasma tissue culture treatmentClear lot number for batch traceabilityIndividually plastic wrappedNon-Pyrogenic, DNase/RNase-FreeHigh performance with 5 growth surfacesCell culture flasks have a growth area of 870cm². Tissue culture treated with a vent cap. Vent caps with 0.22μm hydrophobic filters for gas exchange. Recommended medium volume: 100-150mL.