Learn More
MOLECULAR DEPOT LLC 5M Betaine Solution - 5/2 mL
Catalog No. 5000013419
Encompass_Preferred
5M Betaine Solution PCR Grade consist of a 5M stock solution of betaine (also known as trimethylglycine or N N N triethylammonium acetate) which is an analog of glycine with three methyl groups Betaine is useful in PCR applications because it has an effect on the melting temperature difference between GC and AT base pairs in DNA For in vitro research use only
Catalog No. 50-000-13419 Supplier MOLECULAR DEPOT LLC Supplier No. B2010141
5M Betaine Solution PCR Grade consist of a 5M stock solution of betaine (also known as trimethylglycine or N N N triethylammonium acetate) which is an analog of glycine with three methyl groups Betaine is useful in PCR applications because it has an effect on the melting temperature difference between GC and AT base pairs in DNA For in vitro research use only