Learn More
Ibidi USA Silicone Tubing 0.5 mm ID
SDP
Catalog No. NC0722449
Encompass
0.5 mm ID biocompatible silicone tubing used for a range of flow applications. silicone, sterile, autoclavable, 5 meters
This item is not returnable. View return policy
Catalog No. NC0722449 Supplier Ibidi USA Supplier No. 10840
0.5 mm ID biocompatible silicone tubing used for a range of flow applications. silicone, sterile, autoclavable, 5 meters