Learn More
Teknova 5M Sodium Chloride Solution. 1000mL, Sterile.
SDP
Catalog No. 50843067
Encompass
Sodium chloride is an essential ingredient in the lab. Not only does it help regulate the pH of solutions, but it also prevents growth of bacteria and other contaminants. In addition, salt is a powerful desiccant, meaning that it can help to dry out wet samples. As a result, salt is often used in a wide variety of laboratory procedures.
This item is not returnable. View return policy
Catalog No. 50-843-067 Supplier Teknova Supplier No. S0250
Sodium chloride is an essential ingredient in the lab. Not only does it help regulate the pH of solutions, but it also prevents growth of bacteria and other contaminants. In addition, salt is a powerful desiccant, meaning that it can help to dry out wet samples. As a result, salt is often used in a wide variety of laboratory procedures.