Learn More
Teknova 5M Sodium Hydroxide, Endotoxin Tested, 1000mL, Sterile.
SDP
Catalog No. NC2003602
Encompass
Sodium hydroxide, also known as lye, is a caustic substance that is commonly used as a base in labs for neutralizing acids or creating an alkaline environment for reactions. It is available in a variety of concentrations based on application needs.
This item is not returnable. View return policy
Catalog No. NC2003602 Supplier Teknova Supplier No. S5005
Sodium hydroxide, also known as lye, is a caustic substance that is commonly used as a base in labs for neutralizing acids or creating an alkaline environment for reactions. It is available in a variety of concentrations based on application needs.