Learn More
Epigentek Inc Epigenase 5mC-Hydroxylase TET Activity/Inhibition Assay Kit (Fluorometric) (96 assays)
SDP
Catalog No. 501096327
Encompass_Preferred
The Epigenase 5mC-Hydroxylase TET Activity/Inhibition Assay Kit (Fluorometric) is a complete set of optimized buffers and reagents for measuring activity/inhibition of total cytosine oxygenase TET enzymes in nuclear extracts or purified TET isoforms (TETs 1-3) from a broad range of species such as mammals, plants, fungi, and bacteria, and in a variety of forms including, but not limited to cultured cells and fresh and frozen tissues.
Catalog No. 50-109-6327 Supplier Epigentek Inc Supplier No. P308796
The Epigenase 5mC-Hydroxylase TET Activity/Inhibition Assay Kit (Fluorometric) is a complete set of optimized buffers and reagents for measuring activity/inhibition of total cytosine oxygenase TET enzymes in nuclear extracts or purified TET isoforms (TETs 1-3) from a broad range of species such as mammals, plants, fungi, and bacteria, and in a variety of forms including, but not limited to cultured cells and fresh and frozen tissues.