Learn More
Enzo Life Sciences AMPINEXT™ 5-mC RNA Bisulfite-Seq Easy Kit (Illumina) (12Reactions)
SDP
Catalog No. 502013800
Encompass_Preferred
The AMPINEXT™ 5-mC RNA Bisulfite-Seq Easy Kit (Illumina) is a complete set of optimized reagents designed for easily carrying out RNA bisulfite conversion, followed by a "post-bisulfite" library preparation process for Illumina platform-based bisulfite sequencing, all in one kit. Intended applications include whole transcriptome RNA bisulfite sequencing and various other RNA bisulfite-based next generation sequencing techniques for 5-methylcytosine (5-mC) based RNA methylation analysis. The optimized protocol and components of the kit allow the RNA to be bisulfite converted and fragmented simultaneously, followed by quick non-barcoded (singleplexed) and barcoded (multiplexed) library construction using low-nanogram quantities of bisulfite converted RNA. Assay Time: 6 hours.
Catalog No. 50-201-3800 Supplier Enzo Life Sciences Supplier No. ENZGEN5110012
The AMPINEXT™ 5-mC RNA Bisulfite-Seq Easy Kit (Illumina) is a complete set of optimized reagents designed for easily carrying out RNA bisulfite conversion, followed by a "post-bisulfite" library preparation process for Illumina platform-based bisulfite sequencing, all in one kit. Intended applications include whole transcriptome RNA bisulfite sequencing and various other RNA bisulfite-based next generation sequencing techniques for 5-methylcytosine (5-mC) based RNA methylation analysis. The optimized protocol and components of the kit allow the RNA to be bisulfite converted and fragmented simultaneously, followed by quick non-barcoded (singleplexed) and barcoded (multiplexed) library construction using low-nanogram quantities of bisulfite converted RNA. Assay Time: 6 hours.