Learn More
Cayman Chemical 5-methoxy Tryptamine (hydrochloride), Purity: ≥98%, CAS: 66-83-1, 10 mg
SDP
Catalog No. 502913883
Encompass_Preferred
An analytical reference standard categorized as a tryptamine; intended for research and forensic applications
Catalog No. 50-291-3883 Supplier Cayman Chemical Supplier No. 3678410 MG
An analytical reference standard categorized as a tryptamine; intended for research and forensic applications