Learn More
Cayman Chemical 5-Methoxycarbonylamino-N-acetyltryptamine, Purity: ≥95%, CAS: 190277-13-5, 10 mg
SDP
Catalog No. 502919180
Encompass_Preferred
A partial agonist of MT1 and MT2; inhibits forskolin-induced production of cAMP in CHO cells expressing human MT1 or MT2 (EC50s = 440 and 98 nM, respectively); ocular administration of 5-MCA-NAT reduces intraocular pressure in control and DBA/2J glaucomatous mice
Catalog No. 50-291-9180 Supplier Cayman Chemical Supplier No. 3955310 MG
A partial agonist of MT1 and MT2; inhibits forskolin-induced production of cAMP in CHO cells expressing human MT1 or MT2 (EC50s = 440 and 98 nM, respectively); ocular administration of 5-MCA-NAT reduces intraocular pressure in control and DBA/2J glaucomatous mice