Learn More
Cayman Chemical 5-Methoxyindole-3-acetic Acid, Purity: ≥95%, CAS: 3471-31-6, 500 mg
SDP
Catalog No. 502953796
Encompass_Preferred
A methoxyindole; synthesized in rat pineal gland, retina, and harderian gland; following conversion to a peroxyl radical by HRP, increases the rate of formation of TBARS in liposomes and is cytotoxic to V79 hamster cells; prolongs the estrous cycle and increases diestrous smears, uterine weight, the number of uterine follicles, and plasma levels of 17β-estradiol in female rats
Catalog No. 50-295-3796 Supplier Cayman Chemical Supplier No. 14019500 MG
A methoxyindole; synthesized in rat pineal gland, retina, and harderian gland; following conversion to a peroxyl radical by HRP, increases the rate of formation of TBARS in liposomes and is cytotoxic to V79 hamster cells; prolongs the estrous cycle and increases diestrous smears, uterine weight, the number of uterine follicles, and plasma levels of 17β-estradiol in female rats