Learn More
Cayman Chemical 5methyl2HOBAhydrochlorId 10mg
SDP
Catalog No. 502934992
Encompass_Preferred
An isoketal scavenger; reduces angiotensin II-induced increases in systolic blood pressure in mice
Catalog No. 50-293-4992 Supplier Cayman Chemical Supplier No. 2535110 MG
An isoketal scavenger; reduces angiotensin II-induced increases in systolic blood pressure in mice