Learn More
Cayman Chemical 5methyl2HOBAhydrochlorId 50mg
SDP
Catalog No. 502933104
Encompass_Preferred
An isoketal scavenger; reduces angiotensin II-induced increases in systolic blood pressure in mice
Catalog No. 50-293-3104 Supplier Cayman Chemical Supplier No. 2535150 MG
An isoketal scavenger; reduces angiotensin II-induced increases in systolic blood pressure in mice