Learn More
Cayman Chemical 5-methyl-2'-O-Methylcytidine, Purity: ≥98%, CAS: 113886-70-7, 250 mg
SDP
Catalog No. 502916823
Encompass_Preferred
A modified nucleoside; is found in Huh7 cells infected with Zika virus, HCV, poliovirus, or HIV-1 provirus but not uninfected Huh7 cells; inhibits IL-12 and IL-6 release induced by a TLR9 agonist in mouse spleen cell cultures at 4 µg/ml
Catalog No. 50-291-6823 Supplier Cayman Chemical Supplier No. 37908250 MG
A modified nucleoside; is found in Huh7 cells infected with Zika virus, HCV, poliovirus, or HIV-1 provirus but not uninfected Huh7 cells; inhibits IL-12 and IL-6 release induced by a TLR9 agonist in mouse spleen cell cultures at 4 µg/ml