Learn More
Bio X Cell Polyclonal Human IgG, Purity 95, 5 MG
Supplier: Bio X Cell BE0092R005MG
This polyclonal human IgG is purified from human serum, serving as an ideal non-reactive control for human IgG antibodies in most in vivo and in vitro applications.
- Purified from human serum
- Unconjugated
- Isotype: Human IgG
- Purity ≥95% by SDS-PAGE
- Endotoxin ≤1EU/mg by LAL assay
- Formulated in PBS, pH 7.0, without stabilizers or preservatives
- Molecular weight 150 kDa
- Store at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.