Learn More
Cayman Chemical Panobinostat, 404950-80-7, 5 mg
SDP
Supplier:  Cayman Chemical 132805
Encompass_Preferred
Molecular Formula C21H23N3O2, Purity ≥98%, Formula Weight 349.4, Synonyms: LBH-589; A potent inhibitor of all HDACs, with Ki values ranging from 0.6 to 31 nM for HDAC1-11; leads to acetylation of a range of cellular proteins, resulting in cell cycle arrest and apoptosis in cancer cells; has potential applications in spinal muscular atrophy and HIV therapy.
Catalog No. NC0732649