Learn More
Andwin Scientific 5MG VL MG132 PROTEASOME INHIBI
SDP
Catalog No. NC9326288
Encompass_Preferred
MG-132 Proteasome Inhibitor, 5mg, White Solid. Carbobenzoxy-l-leucyl-l-leucyl-l-leucinal z-leu-leu-leu-cho. Purity> /=90 by HPLC. Soluble in DMSO, CAS No.: 133407-82-6, MW: 475.6. Potent reversible cell-permeable, 1/EA
Catalog No. NC9326288 Supplier Andwin Scientific Supplier No. 4747905
Add to Cart
Add to Cart
MG-132 Proteasome Inhibitor, 5mg, White Solid. Carbobenzoxy-l-leucyl-l-leucyl-l-leucinal z-leu-leu-leu-cho. Purity> /=90 by HPLC. Soluble in DMSO, CAS No.: 133407-82-6, MW: 475.6. Potent reversible cell-permeable, 1/EA