Learn More
Grainger DEVCON Epoxy Adhesive, 5 Minute Epoxy
Catalog No. NC2602170
Encompass_Preferred
Devcon 5 Minute Epoxy is a two-part adhesive that provides a strong, long-lasting bond in just 5 minutes. It is ideal for bonding metal, wood, ceramic, glass, rubber, and many plastics. This epoxy features a 1:1 mix ratio by volume for accurate mixing. It has a working time of 4-5 minutes and reaches handling strength in 20-25 minutes at 75°F. The cured epoxy can withstand temperatures up to 300°F. This 2 oz. syringe contains sufficient material for approximately 30 cubic inches of repair. Devcon 5 Minute Epoxy bonds strongly, sets quickly, and is easy to use.
Catalog No. NC2602170 Supplier Grainger Supplier No. 5A462
Devcon 5 Minute Epoxy is a two-part adhesive that provides a strong, long-lasting bond in just 5 minutes. It is ideal for bonding metal, wood, ceramic, glass, rubber, and many plastics. This epoxy features a 1:1 mix ratio by volume for accurate mixing. It has a working time of 4-5 minutes and reaches handling strength in 20-25 minutes at 75°F. The cured epoxy can withstand temperatures up to 300°F. This 2 oz. syringe contains sufficient material for approximately 30 cubic inches of repair. Devcon 5 Minute Epoxy bonds strongly, sets quickly, and is easy to use.