Learn More
Beckman Coulter Vacutainer/Hemolyse/RIA Tube Adapter, 13.0 MM Diameter, 5 mL, 2/pk
Catalog No. NC1343721
Encompass
Adapter for tubes in Beckman Coulter rotors. Used in benchtop and microcentrifuge rotors: SX4250 SX4400 Color: beige. Quantity of two.
Catalog No. NC1343721 Supplier Beckman Coulter Supplier No. 368467
Adapter for tubes in Beckman Coulter rotors. Used in benchtop and microcentrifuge rotors: SX4250 SX4400 Color: beige. Quantity of two.