Learn More
Ace Glass, Inc. 5ML 14/10 VIAL 5ML 14/10 VIAL
SDP
Catalog No. 50980086
Encompass_Preferred
5mL microscale, thin-wall reaction vial with 14/10 outer joint externally threaded at the opening to allow an o-ring-cap-seal ground glass connection. This thin-walled vial has better heat transfer for the more demanding research project. Supplied with one threaded cap with a hole, one PTFE-faced silicone rubber septum, and one o-ring.
Catalog No. 50-980-086 Supplier Ace Glass, Inc. Supplier No. 959121
5mL microscale, thin-wall reaction vial with 14/10 outer joint externally threaded at the opening to allow an o-ring-cap-seal ground glass connection. This thin-walled vial has better heat transfer for the more demanding research project. Supplied with one threaded cap with a hole, one PTFE-faced silicone rubber septum, and one o-ring.