Learn More
S2 Media 5mL 5x Rappaport-Vassiliadis R10 Broth (5x RV R10), in 60mL Conical Tube, 50/box
Catalog No. 502995771
Encompass_Preferred
5x Rappaport-Vassiliadis R10 Broth (5x RV R10) is a selective enrichment broth used for Salmonella recovery from samples of sanitary importance. This medium selectively enriches for Salmonella spp. because bacteria are typically susceptible to or inhibited by malachite green, high osmotic pressure, and/or low pH. S. typhi and S. choleraesuis are sensitive to malachite green and may be inhibited.• Product meets APHA, BAM, CLSI, ISO, SM-Dairy, SMWW, USDA, and USP-NF reference methods and performance criteria.• RV R10 Broth should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.• Product is a 5x strength.
This item is not returnable. View return policy
Catalog No. 50-299-5771 Supplier S2 Media Supplier No. 5289
5x Rappaport-Vassiliadis R10 Broth (5x RV R10) is a selective enrichment broth used for Salmonella recovery from samples of sanitary importance. This medium selectively enriches for Salmonella spp. because bacteria are typically susceptible to or inhibited by malachite green, high osmotic pressure, and/or low pH. S. typhi and S. choleraesuis are sensitive to malachite green and may be inhibited.• Product meets APHA, BAM, CLSI, ISO, SM-Dairy, SMWW, USDA, and USP-NF reference methods and performance criteria.• RV R10 Broth should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.• Product is a 5x strength.