Learn More
Chemglass Life Sciences 5mL Amber Graduated High Recovery Vial with 20-400 Black Phenolic Open Top Cap and Gray PTFE/Silicone Septa, 20mm OD x 61mm Height
SDP
Catalog No. 5000980621
Encompass_Preferred
Vials are manufactured from Type 1, Class A Amber Borosilicate glass and come with an open-top cap with PTFE Silicone septa.
Catalog No. 50-009-80621 Supplier Chemglass Life Sciences Supplier No. CG4913A005
Vials are manufactured from Type 1, Class A Amber Borosilicate glass and come with an open-top cap with PTFE Silicone septa.