Learn More
Chemglass Life Sciences 5mL Ampul, Drying, Airfree, Schlenk, Round Bottom, 17 O.D. x 147mm Height
SDP
Catalog No. AF008401
Encompass_Preferred
5mL Ampul, Drying, Airfree, Schlenk, Round Bottom, 17 O.D. x 147mm Height. Round bottom, cylindrical drying ampuls with 10mm OD top stem. Manufactured from Type 1 borosilicate glass. Please note: Ampuls are available with top stems to fit AF-0067 Cajon Unions.
Catalog No. AF008401 Supplier Chemglass Life Sciences Supplier No. AF008401
5mL Ampul, Drying, Airfree, Schlenk, Round Bottom, 17 O.D. x 147mm Height. Round bottom, cylindrical drying ampuls with 10mm OD top stem. Manufactured from Type 1 borosilicate glass. Please note: Ampuls are available with top stems to fit AF-0067 Cajon Unions.