Learn More
CELLTREAT Scientific Products 5mL Aspirating Pipet, Short, Individually Wrapped, Sterile
SDP
Catalog No. NC3796184
Encompass_Preferred
Ideal for aspiration of media and sample waste removal without needing to measure the volumeAspirating pipets are supplied without graduations or plugsCompact design of short aspirating pipet provides ergonomic benefit and reduces storage spaceShorter length helps prevent touching pipet tip to surfaces in the hood"
Catalog No. NC3796184 Supplier CELLTREAT Scientific Products Supplier No. 229236
Ideal for aspiration of media and sample waste removal without needing to measure the volumeAspirating pipets are supplied without graduations or plugsCompact design of short aspirating pipet provides ergonomic benefit and reduces storage spaceShorter length helps prevent touching pipet tip to surfaces in the hood"