Learn More
Thomas Scientific 5mL Aspirating Pipet, Individually Wrapped, Sterile
SDP
Catalog No. 501461459
Encompass_Preferred
CELLTREAT offers disposable Aspirating Pipets in 1ml, 2ml and 5ml sizes. Aspirating pipets are supplied without graduations or plugs and are available individually wrapped or bulk-packed. CELLTREAT Scientific Products include a comprehensive "Quality Certificate" with every case of product. The CELLTREAT "Quality Certificate" includes the part number, lot number, sterility assurance level and dosage, endotoxin/pyrogen-free assurance, non-cytotoxicity assurance and RNase-/DNase-free assurance.
Catalog No. 50-146-1459 Supplier Thomas Scientific Supplier No. 1222B09
CELLTREAT offers disposable Aspirating Pipets in 1ml, 2ml and 5ml sizes. Aspirating pipets are supplied without graduations or plugs and are available individually wrapped or bulk-packed. CELLTREAT Scientific Products include a comprehensive "Quality Certificate" with every case of product. The CELLTREAT "Quality Certificate" includes the part number, lot number, sterility assurance level and dosage, endotoxin/pyrogen-free assurance, non-cytotoxicity assurance and RNase-/DNase-free assurance.