Learn More
Andwin Scientific Beaker Cups, Polystyrene B5-1000, Case of 1000
SDP
Catalog No. NC0763039
Encompass_Preferred
The Beaker Cups can be utilized for automated procedures. This product can also perform as an economical, general-purpose sample cup. The disposable beaker cups have a volume capacity of 5 mL and are recommended to use for flame photometry analysis.
Catalog No. NC0763039 Supplier Andwin Scientific Supplier No. 13915985
The Beaker Cups can be utilized for automated procedures. This product can also perform as an economical, general-purpose sample cup. The disposable beaker cups have a volume capacity of 5 mL and are recommended to use for flame photometry analysis.