Learn More
Stellar Scientific CentriCutie® 5mL centrifuge tubes, Sterile, RNase/DNase free, Foam racked, 500/CS
SDP
Catalog No. NC1112695
Encompass
CentriCutie(TM) 5mL centrifuge tubes, sterile, foam racked, 500/CS. These are Very popular in the CRO and CRM community where samples are prepared in larger volumes than the typical research labOther important features are: 20mm (OD) caps that seal smoothly and completely with a single 360° turn. Safe for use from -86°C to 110°C - AutoclavableUses 60% less storage space than 15mL tubesThe 16mm tube diameter is identical to a 15ml tube. NO NEED to purchase additional equipment or adaptersReduced pipette-shaft contamination concernsMade from low-binding virgin polypropylene and are certified RNase & DNase freeSterile tubes are also pyrogen freePerform high speed spins up to 25,000 xg (when fully supported) 4,000xg when suspended in rotor Dual graduations at every .2 and .5mL Leakproof "one turn" capHeight without cap/rim: 54mmHeight with cap: 64mmIts the tube that is good for the planet, and good for your lab!
Catalog No. NC1112695 Supplier Stellar Scientific Supplier No. T05QT
CentriCutie(TM) 5mL centrifuge tubes, sterile, foam racked, 500/CS. These are Very popular in the CRO and CRM community where samples are prepared in larger volumes than the typical research labOther important features are: 20mm (OD) caps that seal smoothly and completely with a single 360° turn. Safe for use from -86°C to 110°C - AutoclavableUses 60% less storage space than 15mL tubesThe 16mm tube diameter is identical to a 15ml tube. NO NEED to purchase additional equipment or adaptersReduced pipette-shaft contamination concernsMade from low-binding virgin polypropylene and are certified RNase & DNase freeSterile tubes are also pyrogen freePerform high speed spins up to 25,000 xg (when fully supported) 4,000xg when suspended in rotor Dual graduations at every .2 and .5mL Leakproof "one turn" capHeight without cap/rim: 54mmHeight with cap: 64mmIts the tube that is good for the planet, and good for your lab!