Learn More
Chemglass Life Sciences 5 mL Centrifuge Tube, Clear, Round Bottom, Snap Cap, Sterile, Nested (200/pack)
SDP
Catalog No. 502733413
Encompass_Preferred
These sterile 5 mL centrifuge tubes are made from USP VI grade polypropylene to provide reliable sample handling for centrifugation, storage, and transport. The tubes feature a round bottom and a snap cap with a secure locking design, are DNase/RNase free, and are supplied nested for sterile workflows.
  • Made from USP VI grade polypropylene, free from heavy metals.
  • Sterile and DNase/RNase free for contamination-sensitive applications.
  • Snap cap with secure lock design for reliable sealing and easy access.
  • Round bottom compatible with high-speed centrifugation up to 30,000 x g.
  • Endotoxin level <0.1 EU.
  • Temperature range -80 °C to +121 °C; autoclavable at 121 °C (15 psi) for 10-15 minutes with proper handling.
  • Supplied nested, 200 per pack, 2,000 per case for sterile handling and efficient storage.
Catalog No. 50-273-3413 Supplier Chemglass Life Sciences Supplier No. CGN430005S
These sterile 5 mL centrifuge tubes are made from USP VI grade polypropylene to provide reliable sample handling for centrifugation, storage, and transport. The tubes feature a round bottom and a snap cap with a secure locking design, are DNase/RNase free, and are supplied nested for sterile workflows.
  • Made from USP VI grade polypropylene, free from heavy metals.
  • Sterile and DNase/RNase free for contamination-sensitive applications.
  • Snap cap with secure lock design for reliable sealing and easy access.
  • Round bottom compatible with high-speed centrifugation up to 30,000 x g.
  • Endotoxin level <0.1 EU.
  • Temperature range -80 °C to +121 °C; autoclavable at 121 °C (15 psi) for 10-15 minutes with proper handling.
  • Supplied nested, 200 per pack, 2,000 per case for sterile handling and efficient storage.