Learn More
Chemglass Life Sciences Chemglass 5 mL Centrifuge Tube, Clear, Round Bottom, Snap Cap, Non-Sterile (Nest)
SDP
Catalog No. 502733414
Encompass_Preferred
Clear 5 mL, round-bottom centrifuge tubes made from USP VI grade polypropylene designed for high-speed centrifugation and reliable sample storage. Tubes feature a frosted marking area and a secure snap cap; they are DNase/RNase free, autoclavable, and packaged in nested packs for laboratory convenience.
  • Made from USP VI grade polypropylene and free from heavy metals.
  • 5 mL capacity with clear graduations and frosted marking area.
  • Secure snap cap for reliable sealing and easy access.
  • Non-sterile, supplied in nested packs (200 per pack, 2,000 per case).
  • Autoclavable at 121°C (250°F) at 15 psi for 10-15 minutes.
  • Temperature range -80°C to +121°C.
  • Rated to 30,000 x g for high-speed centrifugation.
  • DNase/RNase free.
Catalog No. 50-273-3414 Supplier Chemglass Life Sciences Supplier No. CGN430005R
Clear 5 mL, round-bottom centrifuge tubes made from USP VI grade polypropylene designed for high-speed centrifugation and reliable sample storage. Tubes feature a frosted marking area and a secure snap cap; they are DNase/RNase free, autoclavable, and packaged in nested packs for laboratory convenience.
  • Made from USP VI grade polypropylene and free from heavy metals.
  • 5 mL capacity with clear graduations and frosted marking area.
  • Secure snap cap for reliable sealing and easy access.
  • Non-sterile, supplied in nested packs (200 per pack, 2,000 per case).
  • Autoclavable at 121°C (250°F) at 15 psi for 10-15 minutes.
  • Temperature range -80°C to +121°C.
  • Rated to 30,000 x g for high-speed centrifugation.
  • DNase/RNase free.