Learn More
Waters Corp Cryotube Domino, 5 mL, 24-Tube Capacity, Machine-Readable
Catalog No. NC3597591 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The 5mL Cryotube Domino is designed to hold a variety of cryotubes, with a capacity for up to 24. These machine-readable holders interface with the Andrew+ pipetting robot, automating laboratory workflows. Equipped with QR and ARUCO codes, they are essential for automation configurations.
  • Designed to hold a growing array of cryotubes.
  • Holds up to 24 tubes.
  • Functions as a machine-readable consumable holder.
  • Creates the interface between various consumable types and the Andrew+ pipetting robot.
  • Enables seamless automation of many laboratory workflows.
  • Equipped with QR codes for critical labware information.
  • Includes ARUCO codes for spatial positioning.
  • Indispensable part of any automation configuration.
Catalog No. NC3597591 Supplier Waters Corp Supplier No. 186010575
The 5mL Cryotube Domino is designed to hold a variety of cryotubes, with a capacity for up to 24. These machine-readable holders interface with the Andrew+ pipetting robot, automating laboratory workflows. Equipped with QR and ARUCO codes, they are essential for automation configurations.
  • Designed to hold a growing array of cryotubes.
  • Holds up to 24 tubes.
  • Functions as a machine-readable consumable holder.
  • Creates the interface between various consumable types and the Andrew+ pipetting robot.
  • Enables seamless automation of many laboratory workflows.
  • Equipped with QR codes for critical labware information.
  • Includes ARUCO codes for spatial positioning.
  • Indispensable part of any automation configuration.