Learn More
Bellco Glass, Inc. 5mL Culture Tube and Dual Cap, PP, Sterile
SDP
Catalog No. 502330682
Encompass_Preferred
Dual caps allow for either vented or sealed positions14mL tubes feature molded graduationsPolystyrene tubes are optically clear to visualize culturesPolypropylene tubes offer better chemical resistance Pack Description25/Re-Sealable BagCase Quantity500SterileYesNon-PyrogenicYesRNase/DNase FreeYesAutoclavableYesPrimary ColorTube: NaturalSecondary ColorCap: NaturalPrimary MaterialTube: PolypropyleneSecondary MaterialCap: LDPEMaximum Volume5mLGraduatedNoMax Force (RCF) g3000 x g
Catalog No. 50-233-0682 Supplier Bellco Glass, Inc. Supplier No. 230433000
Dual caps allow for either vented or sealed positions14mL tubes feature molded graduationsPolystyrene tubes are optically clear to visualize culturesPolypropylene tubes offer better chemical resistance Pack Description25/Re-Sealable BagCase Quantity500SterileYesNon-PyrogenicYesRNase/DNase FreeYesAutoclavableYesPrimary ColorTube: NaturalSecondary ColorCap: NaturalPrimary MaterialTube: PolypropyleneSecondary MaterialCap: LDPEMaximum Volume5mLGraduatedNoMax Force (RCF) g3000 x g