Learn More
Bellco Glass, Inc. 5mL Culture Tube and Dual Cap, PS, Sterile
SDP
Catalog No. 502330475
Encompass_Preferred
Dual caps allow for either vented or sealed positions14mL tubes feature molded graduationsPolystyrene tubes are optically clear to visualize cultures Pack Description25/Re-Sealable BagCase Quantity500SterileYesNon-PyrogenicYesRNase/DNase FreeYesAutoclavableNoPrimary ColorTube: ClearSecondary ColorCap: NaturalPrimary MaterialTube: PolystyreneSecondary MaterialCap: LDPEMaximum Volume5mLGraduatedNoGraduation Increments (Subdivisions)NAMax Force (RCF) g1400 xg
Catalog No. 50-233-0475 Supplier Bellco Glass, Inc. Supplier No. 230434000
Dual caps allow for either vented or sealed positions14mL tubes feature molded graduationsPolystyrene tubes are optically clear to visualize cultures Pack Description25/Re-Sealable BagCase Quantity500SterileYesNon-PyrogenicYesRNase/DNase FreeYesAutoclavableNoPrimary ColorTube: ClearSecondary ColorCap: NaturalPrimary MaterialTube: PolystyreneSecondary MaterialCap: LDPEMaximum Volume5mLGraduatedNoGraduation Increments (Subdivisions)NAMax Force (RCF) g1400 xg