Learn More
S2 Media 5mL Deionized (DI) Water, Sterile, in 50mL PP Conical Tube, 50/box
Catalog No. 502995961
Encompass_Preferred
Deionized (DI) Water created using an ion exchange process. It is purified water lacking ions, such as those from sodium, calcium, and iron. This product is intended for general laboratory use in the preparation of dilutions, suspensions, and test samples for microbial testing. Sterile DI Water is not for preparations intended for parenteral administration. End-users of commercially prepared culture media, diluents, and reagents should perform QC testing in accordance with applicable government regulatory agencies and in compliance with accreditation requirements, as applicable. Water, buffers, and saline products are supplied by S2Media in accordance with our quality systems.• DI Water, Sterile should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
Catalog No. 50-299-5961 Supplier S2 Media Supplier No. 5456
Deionized (DI) Water created using an ion exchange process. It is purified water lacking ions, such as those from sodium, calcium, and iron. This product is intended for general laboratory use in the preparation of dilutions, suspensions, and test samples for microbial testing. Sterile DI Water is not for preparations intended for parenteral administration. End-users of commercially prepared culture media, diluents, and reagents should perform QC testing in accordance with applicable government regulatory agencies and in compliance with accreditation requirements, as applicable. Water, buffers, and saline products are supplied by S2Media in accordance with our quality systems.• DI Water, Sterile should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.