Learn More
Alkali Scientific 5mL False Bottom Analyzer Tubes with Threads, 16×97mm, Polypropylene (PP), Clear, 1000/Pk
SDP
Catalog No. 5000661964
Encompass_Preferred
These 5mL False Bottom Analyzer Tubes (16×97mm) with Threads are designed for use in high volume chemistry and diagnostic analyzers. They are compatible with popular analyzers from Hitachi, Olympus, Corning, Ortho, Kodak, Beckman, IA, Tosoh, Vitros and more. Manufactured from high grade polypropylene (PP), these tubes offer excellent chemical resistance and can be safely autoclaved at 121 °C. With a conical false bottom for reduced dead volume and accurate sample retrieval, these tubes are ideal for serum collection, sample transport, or storage. Available in natural or amber options to protect light sensitive samples, they are self standing and designed to fit most universal analyzer racks, including AU 5000 systems. Features: 5mL nominal capacity, 16×97 mm Compatible with Hitachi, Olympus, Corning, Ortho, Beckman, Vitros, Tosoh, and other analyzers Made of chemical resistant polypropylene; autoclavable at 121 °C Available in natural or amber PP for light sensitific.
Catalog No. 50-006-61964 Supplier Alkali Scientific Supplier No. FBAT05
These 5mL False Bottom Analyzer Tubes (16×97mm) with Threads are designed for use in high volume chemistry and diagnostic analyzers. They are compatible with popular analyzers from Hitachi, Olympus, Corning, Ortho, Kodak, Beckman, IA, Tosoh, Vitros and more. Manufactured from high grade polypropylene (PP), these tubes offer excellent chemical resistance and can be safely autoclaved at 121 °C. With a conical false bottom for reduced dead volume and accurate sample retrieval, these tubes are ideal for serum collection, sample transport, or storage. Available in natural or amber options to protect light sensitive samples, they are self standing and designed to fit most universal analyzer racks, including AU 5000 systems. Features: 5mL nominal capacity, 16×97 mm Compatible with Hitachi, Olympus, Corning, Ortho, Beckman, Vitros, Tosoh, and other analyzers Made of chemical resistant polypropylene; autoclavable at 121 °C Available in natural or amber PP for light sensitific.