Learn More
Ace Glass, Inc. 5ML GAS TIGHT SYRINGE 5Ml Gas Tight Syringe
SDP
Catalog No. 50972630
Encompass
5mL gas tight syringe for sample introduction into solvent delivery systems. PTFE coated plunger with luer lock.
Catalog No. 50-972-630 Supplier Ace Glass, Inc. Supplier No. 593208
5mL gas tight syringe for sample introduction into solvent delivery systems. PTFE coated plunger with luer lock.