Learn More
Olerup Inc SSP® GelRedTM Dropper Bottle 5ML
SDP
Catalog No. NC1139154
Encompass
GelRedTM is a sensitive, stable and environmentally safe fluorescentnucleic acid dye designed to replace the toxic ethidium bromide forstaining dsDNA, ssDNA or RNA in agarose gels or polyacrylamidegels. The dropper bottle is specially designed to exclude damaging UVlight. GelRed™ should be shielded from light and exposure to lightshould be avoided during long-term storage
Catalog No. NC1139154 Supplier Olerup Inc Supplier No. 103.30205
GelRedTM is a sensitive, stable and environmentally safe fluorescentnucleic acid dye designed to replace the toxic ethidium bromide forstaining dsDNA, ssDNA or RNA in agarose gels or polyacrylamidegels. The dropper bottle is specially designed to exclude damaging UVlight. GelRed™ should be shielded from light and exposure to lightshould be avoided during long-term storage