Learn More
Metrohm USA 5ML GLASS CYLINDER COMPLETE
SDP
Supplier:  Metrohm USA 61574150
Encompass
Glass cylinder unit complete with cylinder, piston, cylinder base, valve disk and centering tube. For dosing units.
Catalog No. NC0182304