Learn More
Cole-Parmer PTFE Tissue Grinder, Glass Vessel; 5 mL
Supplier:  Cole-Parmer 4446814
Encompass_Preferred
These tissue grinders are capable of reducing cellular tissue to primary cells. Glass vessels are made of precision borosilicate glass. Grinding chamber clearance is 0.15 to 0.25 mm.
Catalog No. NC1842219