Learn More
Ace Glass, Inc. 5mL hemispherical fabric mantle, Glas-Col, 27mm max flask diameter, 0.66in inside depth, 1.56in OD, 0.91in height, 30vac max, 12 watts, not rated
SDP
Catalog No. 50977262
Encompass
5mL hemispherical fabric Glas-col mantle, adaptable to unusual shapes and sizes, and will withstand 450 Deg.C. Mantle has a separable 2-wire 3 cord with locking connector. cULus rated. CSA certified.
Catalog No. 50-977-262 Supplier Ace Glass, Inc. Supplier No. 1203501
5mL hemispherical fabric Glas-col mantle, adaptable to unusual shapes and sizes, and will withstand 450 Deg.C. Mantle has a separable 2-wire 3 cord with locking connector. cULus rated. CSA certified.