Learn More
Chemglass Life Sciences 5mL Single Neck Round Bottom Flask (RBF) , 19/22 Outer Joint
SDP
Catalog No. CG150681
Encompass_Preferred
5mL Single Neck Round Bottomed Flask, 19/22 Outer Joint. Round bottom, HEAVY WALL single short neck flask with a standard taper outer joint.
Catalog No. CG150681 Supplier Chemglass Life Sciences Supplier No. CG150681
5mL Single Neck Round Bottomed Flask, 19/22 Outer Joint. Round bottom, HEAVY WALL single short neck flask with a standard taper outer joint.