Learn More
CELLTREAT Scientific Products 5mL Low Retention Pipette Tips, Racked, Sterile, 144 Tips (24 tips per rack)
SDP
Catalog No. NC2206336
Encompass_Preferred
Sterile, accurate liquid transfer of larger volumes of media and solutions. Universal fit is compatible with a range of popular pipettes. More precise than serological pipets for volumes of 1 to 10mL
This item is not returnable. View return policy
Catalog No. NC2206336 Supplier CELLTREAT Scientific Products Supplier No. 229039
Sterile, accurate liquid transfer of larger volumes of media and solutions. Universal fit is compatible with a range of popular pipettes. More precise than serological pipets for volumes of 1 to 10mL