Learn More
Bellco Glass, Inc. 5mL Low Retention Pipette Tips, Racked, Sterile
SDP
Catalog No. 502329975
Encompass_Preferred
Sterile, accurate liquid transfer of larger volumes of media and solutionsUniversal fit is compatible with a range of popular pipettesMore precise than serological pipets for volumes of 1 to 10mL5mL tips have molded graduations, with 0.5mL increments Pack Description24 Tips/RackCase Quantity144SterileYesRNase/DNase FreeYesAutoclavableYesLow RetentionYesFilterNoRackedYesPrimary ColorNaturalPrimary MaterialPolypropyleneMaximum Volume5000µLGraduatedYes
Catalog No. 50-232-9975 Supplier Bellco Glass, Inc. Supplier No. 229039000
Sterile, accurate liquid transfer of larger volumes of media and solutionsUniversal fit is compatible with a range of popular pipettesMore precise than serological pipets for volumes of 1 to 10mL5mL tips have molded graduations, with 0.5mL increments Pack Description24 Tips/RackCase Quantity144SterileYesRNase/DNase FreeYesAutoclavableYesLow RetentionYesFilterNoRackedYesPrimary ColorNaturalPrimary MaterialPolypropyleneMaximum Volume5000µLGraduatedYes