Learn More
Trajan Scientific 5ML LURLOCK GAS TGHT SYRINGE
SDP
Catalog No. 502530299
Encompass_Preferred
SGE LC Pump Priming Syringe, Volume: 5 mL, For Use With: Used for loop fill injection or sample loop flush, Model: 5MDF-LL-GT, Needle Point Style: LC, Quantity: Each, Manual Syringe
Catalog No. 50-253-0299 Supplier Trajan Scientific Supplier No. 008762
SGE LC Pump Priming Syringe, Volume: 5 mL, For Use With: Used for loop fill injection or sample loop flush, Model: 5MDF-LL-GT, Needle Point Style: LC, Quantity: Each, Manual Syringe