Learn More
S2 Media 15x100mm MacConkey Agar with 4µg/mL Ceftriaxone, 10/pack
Catalog No. 502995975
Encompass_Preferred
MacConkey Agar with 4µg/mL Ceftriaxone is a selective and differential medium used for the detection and isolation of Gram-negative bacteria that are resistant to ceftriaxone, particularly those producing extended-spectrum β-lactamases (ESBLs) or exhibiting other third-generation cephalosporin resistance mechanisms.• Product base meets AOAC, BAM, CLSI, and Harmonized USP/EP/JP, NF, SM-Dairy, and SMWW reference methods and performance criteria.• MacConkey Agar should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
This item is not returnable. View return policy
Catalog No. 50-299-5975 Supplier S2 Media Supplier No. 5446
MacConkey Agar with 4µg/mL Ceftriaxone is a selective and differential medium used for the detection and isolation of Gram-negative bacteria that are resistant to ceftriaxone, particularly those producing extended-spectrum β-lactamases (ESBLs) or exhibiting other third-generation cephalosporin resistance mechanisms.• Product base meets AOAC, BAM, CLSI, and Harmonized USP/EP/JP, NF, SM-Dairy, and SMWW reference methods and performance criteria.• MacConkey Agar should be used by trained professionals within a laboratory setting. • For in vitro diagnostic (IVD) use.• Product is not intended for use in the diagnosis or treatment of disease or other human conditions.