Learn More
Bellco Glass, Inc. 5mL Macro Centrifuge Tube
SDP
Catalog No. 502329562
Encompass_Preferred
Description5mL Macro Centrifuge Tubes offer the convenience of a micro centrifuge tube with the ability to process sample volumes up to 5mLTube diameter and shape is the same as a standard 15mL conical bottom centrifuge tube, enabling use with existing equipmentTubes have engraved graduationsAttached flat top cap is easy to open and close with one handCenter of the cap includes a thin “membrane” area designed for easy puncture and access by a syringe/needleBlack tubes protect light sensitive compounds and eliminate the need to wrap tubes with aluminum foil Pack Description50/Re-sealable BagCase Quantity100SterileNoAutoclavableYesPrimary ColorBlackPrimary MaterialPolypropyleneMaximum Volume5mLGraduatedYesGraduation Increments (Subdivisions)0.2mLMax Force (RCF)g25000RNase/DNase FreeYes
Catalog No. 50-232-9562 Supplier Bellco Glass, Inc. Supplier No. 229439000
Description5mL Macro Centrifuge Tubes offer the convenience of a micro centrifuge tube with the ability to process sample volumes up to 5mLTube diameter and shape is the same as a standard 15mL conical bottom centrifuge tube, enabling use with existing equipmentTubes have engraved graduationsAttached flat top cap is easy to open and close with one handCenter of the cap includes a thin “membrane” area designed for easy puncture and access by a syringe/needleBlack tubes protect light sensitive compounds and eliminate the need to wrap tubes with aluminum foil Pack Description50/Re-sealable BagCase Quantity100SterileNoAutoclavableYesPrimary ColorBlackPrimary MaterialPolypropyleneMaximum Volume5mLGraduatedYesGraduation Increments (Subdivisions)0.2mLMax Force (RCF)g25000RNase/DNase FreeYes